Learn More
Tocris Bioscience™ Ac-FLTD-CMK
Potent and selective inhibitor of caspases 1, 5 and 4
Brand:  Tocris Bioscience™ 7242/1
Product Code. 18035742
  • 462.00€ / 1mg
Explore more special offers
Description
Description
Ac-FLTD-CMK is a potent and selective inhibitor of caspases 1, 5 and 4 (IC50 values = 46.7 nM, 0.33 μM, 1.49 μM, respectively), and inhibits murine caspase-11 at 10 μM. Ac-FLTD-CMK inhibits cleavage of gasdermin D by caspases, suppresses pyroptosis and reduces IL-1β release in macrophages.
Specifications
Specifications
2376255-48-8
Caspase Inhibitors
0.95
1 mg
C26H37ClN4O8