Learn More
Haleco™ Absorbent Polymer Neutralizing Powder
Product Code. 11937364
Click to view available options
:
White
Product Code. 11937364
Brand: Haleco™ JLN110B
Only null left
Add to basket
Add to basket
compliance-icons
Product Identifier
  • neutrabio powder bucket
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.