Learn More
Thermo Scientific™ ABsolute™ Probe-Based Detection qPCR Master Mixes
Product Code. p-4526932 Shop All Thermo Scientific Products
Click to view available options
Description:
Absolute QPCR ROX (500nM) Mix, QPCR ROX mix; 1.25mL vial; 100 rxns
Quantity:
2 x 1.25mL vials; 100 x 50µL rxns
Unit Size:
200 reactions
Product Code. 11837682
Brand: Thermo Scientific™ AB1138/A
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Thermo Scientific™ ABsolute qPCR Master Mixes offer sensitivity and reproducibility for real-time PCR applications.
Enjoy sensitivity and reproducibility in real-time PCR applications with Thermo Scientific™ ABsolute™ Probe-Based Detection qPCR Master Mixes.
ABsolute qPCR Master Mixes are designed to achieve the most consistent and reproducible data across all QPCR platforms. Proprietary additives give enhanced and consistent end point readings as well as low Ct values. All mixes contain Thermo-Start Taq DNA Polymerase, for increased specificity, and dTTP for increased reaction efficiency and sensitivity (low Ct).
  • Maximum specificity—Chemically-modified Thermo-Start™ Taq remains completely inactive until heated to 95°C for 15 minutes, thereby preventing nonspecific amplification
  • Improved efficiency—The use of dTTP in place of dUTP increases reaction efficiency by approximately 5% and therefore reduces reaction time
  • Increased sensitivity—The combination of Thermo-Start Taq and dTTP increases sensitivity and allows for detection of low copy number templates
  • Instrument specific—Individually optimized qPCR mixes designed for all major qPCR / Real-Time PCR instruments
  • Reproducible results—All mixes undergo extensive quality control testing to ensure batch-to-batch consistency
  • Simple protocol—Master mix format reduces protocol steps and minimizes risk of pipetting errors and sample contamination
Includes:
ABsolute qPCR Buffer (Thermo-Start DNA polymerase, dNTPs, MgCl2, ROX™ passive reference dye (only in ROX mixes); Separate ROX vial (only in kits indicated)
Recommended for:
Ideal for qPCR analysis in molecular biology applications, including gene expression and SNP typing
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage ABsolute qPCR Buffer (Thermo-Start DNA polymerase, dNTPs, MgCl2, ROX™ passive reference dye (only in ROX mixes); Separate ROX vial (only in kits indicated). Store at -20°C. Avoid freeze/thaw cycles
Description Absolute QPCR ROX (500nM) Mix, QPCR ROX mix; 1.25mL vial; 100 rxns
Quantity 2 x 1.25mL vials; 100 x 50µL rxns
For Use With (Application) Gene Expression, Genotyping, miRNA analysis
Activation Time heated to 95°C for 15 minutes
Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser's own internal research. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.