Learn More
Abnova™ 8p Subtelomere (Texas Red) FISH Probe
Labeled FISH probes for identification of subtelomere aberrations using Fluorescent In Situ Hybridization Technique.
Brand:  Abnova™ FE0072.20uL
Product Code. 16052845
  • 938.00€ / 20µL
Explore more special offers
Includes: FISH Hybridization Buffer (80μL)
Description
Description
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
  • Dual colored probes for fast, sensitive, and specific detection
  • Work on metaphase spread, paraffin embedded, and frozen tissue
  • Identify gene amplification, loss, and translocation
  • High signal to noise ratio
  • Low cross reactivity
Specifications
Specifications
Store at 4°C in the dark.
Fluorescent In Situ Hybridization (Cell)
8p Subtelomere
20 μL
Texas Red
FISH Hybridization Buffer (80 uL)
8p Subtelomere(Texas Red) FISH Probe