missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ A 939572
Potent SCD-1 inhibitor
233.26€ - 894.15€
Specifications
Chemical Name or Material | 4-(2-Chlorophenoxy)-N-[3-[(methylamino)carbonyl]phenyl]-1-piperidinecarboxamide |
---|---|
CAS | 1032229-3-6 |
Target | Compounds for Stem Cell Differentiation |
Molecular Formula | C20H22ClN3O3 |
Formula Weight | 387.86 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18716239
|
Tocris Bioscience™
4845/10 |
10 mg |
233.26€
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18721088
|
Tocris Bioscience™
4845/50 |
50 mg |
894.15€
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent stearoyl-CoA desaturase 1 (SCD1) inhibitor (IC50 = 0.4 nM). Selective for SCD1 over a range of kinases and hERG channels. Inhibits proliferation of squamous cell carcinoma FaDu cells in vitro. Also induces cell death of undifferentiated human embryonic stem cells (ESCs). Orally bioavailable.Specifications
4-(2-Chlorophenoxy)-N-[3-[(methylamino)carbonyl]phenyl]-1-piperidinecarboxamide | |
Compounds for Stem Cell Differentiation | |
387.86 |
1032229-3-6 | |
C20H22ClN3O3 | |
>99% |
Safety and Handling
Recommended Storage : Store at 4°C