missing translation for 'onlineSavingsMsg'
Learn More

A 68930 hydrochloride, Tocris Bioscience™

Product Code. 30001264
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent, selective D1-like agonist

Potent and selective D1-like dopamine receptor agonist (EC50 values are 2.1 and 3910 nM for D1-like and D2-like receptors respectively). Centrally active following systemic administration in vivo.

Chemical Identifiers

CAS 130465-39-3
Molecular Formula C16H18ClNO3
Molecular Weight (g/mol) 307.774
InChI Key PQPGUUQPTSMLKU-YYLIZZNMSA-N
Synonym a 68930 hydrochloride, a-68930 hydrochloride, cis-+/--1-aminomethyl-3,4-dihydro-3-phenyl-1h-2-benzopyran-5,6-diol hydrochloride, cis-1-aminomethyl-3-phenylisochroman-5,6-diol hydrochloride, 1r,3s-1-aminomethyl-3-phenyl-3,4-dihydro-1h-2-benzopyran-5,6-diol hydrochloride, a-68930 hydrochloride hplc , powder, cis-1-aminomethyl-3-phenyl-3,4-dihydro-5,6-dihydroxy-1h-2-benzopyran hydrochloride, cis-inverted exclamation marka-1-aminomethyl-3,4-dihydro-3-phenyl-1h-2-benzopyran-5,6-diol hydrochloride
PubChem CID 9904672
IUPAC Name (1R,3S)-1-(aminomethyl)-3-phenyl-3,4-dihydro-1H-isochromene-5,6-diol;hydrochloride
SMILES C1C(OC(C2=C1C(=C(C=C2)O)O)CN)C3=CC=CC=C3.Cl

Specifications

Quantity 10 mg
Formula Weight 307.78
Percent Purity >98%
Chemical Name or Material A 68930 hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'