missing translation for 'onlineSavingsMsg'
Learn More

A 61603 hydrobromide, Tocris Bioscience™

Product Code. 18025462
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

α1A agonist

Potent α-adrenoceptor agonist that is at least 35-fold more potent at α1A than at α1B or α1D sites. Induces dose response increases in spontaneous Ca2+ transients in rat ventricular myocytes in vitro (EC50 = 6.9 nmol/L).

Chemical Identifiers

CAS 107756-30-9
Molecular Formula C14H20BrN3O3S
Molecular Weight (g/mol) 390.296
InChI Key LRFLWCZMTGTUEP-UHFFFAOYSA-N
Synonym a 61603 hydrobromide, n-5-4,5-dihydro-1h-imidazol-2-yl-2-hydroxy-5,6,7,8-tetrahydronaphthalen-1-yl methanesulfonamide hydrobromide
PubChem CID 9865178
IUPAC Name N-[5-(4,5-dihydro-1H-imidazol-2-yl)-2-hydroxy-5,6,7,8-tetrahydronaphthalen-1-yl]methanesulfonamide;hydrobromide
SMILES CS(=O)(=O)NC1=C(C=CC2=C1CCCC2C3=NCCN3)O.Br

Specifications

Quantity 50 mg
Formula Weight 390.29
Percent Purity >98%
Chemical Name or Material A 61603 hydrobromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'