missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ 96-well V-Bottom PCR Microplates
GREENER_CHOICE

Product Code. 10529203 Shop All Eppendorf Products
Click to view available options
Color:
Gray, White Bottom, White Bottom
White, Black Bottom, Black Bottom
White, Clear Bottom
Quantity:
240/Pk.
80/Pk.
Unit Size:
Pack of 240
Pack of 80
This item is not returnable. View return policy

Product Code. 10529203

Brand: Eppendorf™ 0030605306

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Specifications

Certified Yes
Format Standard
Detection Method Quantitative Protein Analysis
Array 8 x 12
Color White, Clear Bottom
Product Type 96-well PCR Microplate
Material Polypropylene
No. of Wells 96
Well Shape Conical Bottom (V-well)
Dnase Free Yes
For Use With (Application) For PCR techniques
Max. Heat Stability 121°C
Quantity 240/Pk.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.