Learn More
Greiner Bio-One 96-well CrystalQuick™ Plus Crystallisation Microplates
For automatised applications. Greiner Bio-One 96-Well CrystalQuick™ Plus Crystallisation Microplates are optimised for sealing with VIEWseal adhesive film.
Brand: Greiner Bio-One 609830
This item is not returnable.
View return policy
Description
- Non-sterile
- Polystyrene
- Good optical properties
- Hydrophobic surface
- With crystallisation well per reservoir
- Alphanumeric well coding
- With CrystalDrop™ lid, simultaneous experiments are possible with the sitting drop and hanging drop method
Specifications
Standard | |
Clear | |
4.1 μL | |
Non-sterile | |
Sitting drop applications with different well profiles and material properties | |
40/Cs. |
8 x 12 | |
Polystyrene | |
96 | |
Square, Flat Bottom (F-well) | |
40 | |
14.4 mm |