missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One 96-well CrystalQuick™ Plus Crystallisation Microplates

Product Code. 10083973
Click to view available options
Volume (Metric) Well:
3.9 μL
4.1 μL
Height (Metric):
3.9 mm
14.4 mm
Unit Size:
Pack of 40
Pack of 80
This item is not returnable. View return policy

Product Code. 10083973

Brand: Greiner Bio-One 609180

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

For automatised applications. Greiner Bio-One 96-Well CrystalQuick™ Plus Crystallisation Microplates are optimised for sealing with VIEWseal adhesive film.

  • Non-sterile
  • Polystyrene
  • Good optical properties
  • Hydrophobic surface
  • With crystallisation well per reservoir
  • Alphanumeric well coding
  • With CrystalDrop™ lid, simultaneous experiments are possible with the sitting drop and hanging drop method
TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Array 8 x 12
Color Clear
Material Polystyrene
Volume (Metric) Well 3.9 μL
No. of Wells 96
Sterility Non-sterile
Well Shape Square, Flat Bottom (F-well)
For Use With (Application) Sitting drop applications with different well profiles and material properties
No. per Case 80
Quantity 80/Cs.
Height (Metric) 3.9 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.