Learn More
Greiner Bio-One 96-well CrystalQuick™ Crystallisation Microplates
For automatised applications. Greiner Bio-One 96-Well CrystalQuick™ Crystallisation Microplates are optimised for sealing with VIEWseal adhesive film.
Brand:  Greiner Bio-One 609101
 View more versions of this product
Product Code. 10010372
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • Non-sterile
  • Polystyrene
  • Good optical properties
  • With crystallisation well per reservoir
  • Alphanumeric well coding
  • With CrystalDrop™ lid, experiments are possible with sitting drop and hanging drop metho
    TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
Clear
Polystyrene
96
Square, Flat Bottom (F-well)
40
14.4 mm
8 x 12
Standard
4.1 μL
Non-sterile
Sitting drop applications with different well profiles and material properties
40/Cs.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products