Learn More
uvex i-5 Safety Glasses with CBR65 Lens
Click to view available options
Lens Tint:
Clear
Gray
Description
- Sophisticated features ensure superior user comfort when worn for extended periods of time
- Soft, flexible nosepiece for enhanced comfort and a non-slip fit
- X-tended eyeshield protects against dust, dirt, and flying particles
- X-tended sideshield increases protection without restricting peripheral vision
- 3-position inclination for a personalized fit
- Extendable x-twist side arm technology offers flexible & convenient adjustment
- Soft ear pieces for a pressure-free, non-slip fit
- Reliable UV 400 protection
Supravision Excellence Coating
- Inside: Anti-fog
- Outside: Scratch-resistant and chemical-resistant
Specifications
Specifications
Certifications/Compliance | EN 166:2001, EN 172 |
Lens Type | Supravision Excellence |
Lens Coating | Anti-fog |
Lens Tint | Gray |
Product Type | Ergonomic Safety Glasses |
Frame Color | Anthracite, Lime |
Model | 9183 i-5 |
Transmittance Value | 0.23 |
Material | Polycarbonate |
Disposable | Reusable |