missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ 900 mm ReVo™ Class II, Type A2 Microbiological Safety Cabinet
ReVo™ Class II, Type A2 Microbiological Safety Cabinets provide personnel, product and environmental protection from biohazardous agents and hazardous particulates.
12575.00€ - 13565.00€
Specifications
Certifications/Compliance | EN 12469 Certified by TÜV |
---|---|
Chamber Material | Stainless Steel (304) |
Product Type | Class II Biosafety Cabinet |
Noise Level | 43 dBA |
Outlets | 2 |
Product Code | Brand | Ports | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Ports | Includes | Price | Quantity & Availability | ||||
17810754
|
Labconco™
3R2310031 |
Schuko Plug | Base Stand Not Included |
12575.00€
1 unit |
Estimated Shipment: 06-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17840754
|
Labconco™
3R2310131 |
Schuko Plug | Base Stand |
13565.00€
1 unit |
Estimated Shipment: 06-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Provide personnel, product and environmental protection from biohazardous agents and hazardous particulates.
- Other appropriate applications include work with antineoplastic drugs, genetic material, and additional substances that generate hazardous airborne particulates..
- All models are EN 12469 compliant with additional certification by TÜV Nord.
- These biological safety cabinets are designed to discharge HEPA-filtered exhaust air directly into the laboratory, or into an exhaust system through the optional Canopy Connection.
- When canopy-connected, the biosafety cabinets may be used for applications involving minute quantities of volatile toxic chemicals and tracer amounts of radionuclides as an adjunct to microbiological research.
- During operation, room air is drawn into the inlet grill.
- The air in the plenum beneath the work surface is a mixture of unfiltered room air and air that has just passed through the work area.
- This contaminated air is drawn by the blower through the back plenum of the cabinet, where approximately 70% of the air is recirculated through the supply HEPA filter and back over the work area.
- The balance of the contaminated air is discharged to the environment after passing through the exhaust HEPA filter.
Specifications
EN 12469 Certified by TÜV | |
Class II Biosafety Cabinet | |
2 | |
200 mm | |
H14 HEPA | |
797 x 1004 x 1350 mm |
Stainless Steel (304) | |
43 dBA | |
85 W | |
HEPA Exhaust Filter | |
7.87 in. | |
230 V |