Learn More
Mitutoyo™ 811 Series 20 to 90 Shore D Analogue Display Durometers
Brand: Mitutoyo™ 811333
This item is not returnable.
View return policy
Specifications
Analog | |
Hard elastomers, plastics, hard rubber, ebonite, etc. | |
20 to 90 Shore D |