Learn More
Rogo Sampaic™ Stainless Steel 8-Piece Dissecting Set
A convenient and comprehensive set of high-quality dissection instruments provided in a handsome black, red-lined zippered case. Rogo Sampaic™ Stainless Steel 8-Piece Dissecting Sets are excellent for disection, surgical and general laboratory needs.
Brand: Rogo Sampaic™ FMQ011
Description
- 1 pair of strong, sharp/round 140mm scissors
- 1 pair of Iris 110mm scissors
- 1 pair of strong rounded 140mm pliers
- 1 pair of find rounded 110mm tweezers/forceps
- 1 No. 4 scalpel handle
- 1 sachet of 5 scalpel blades, No. 23
- 2 straight-pointed needles for dissociating stitches
Specifications
1 Ea. | |
Dissecting Kit, 8 Piece |