missing translation for 'onlineSavingsMsg'
Learn More

8-Bromoquinoline, 98%, Thermo Scientific Chemicals

Product Code. 15483327
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16567-18-3
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.06
MDL Number MFCD00191859
InChI Key PIWNKSHCLTZKSZ-UHFFFAOYSA-N
Synonym quinoline, 8-bromo, 8-bromo-quinoline, 8-bromoquinoline, pubchem5816, 8-bromquinoline, acmc-209dt3, ksc181q5t, buttpark 33\04-57
PubChem CID 140109
IUPAC Name 8-bromoquinoline
SMILES BrC1=C2N=CC=CC2=CC=C1

Specifications

Density 1.594
Boiling Point 302°C to 304°C
Flash Point >110°C (230°F)
Refractive Index 1.6725
Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 208.06
Percent Purity 98%
Chemical Name or Material 8-Bromoquinoline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'