missing translation for 'onlineSavingsMsg'
Learn More

7-Fluorochroman-4-one, may contain up to 15% 5-isomer, 98% (sum of isomers), Thermo Scientific™

Product Code. 15445135
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15445135

Brand: Thermo Scientific Alfa Aesar H31881.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113209-68-0
Molecular Formula C9H7FO2
Molecular Weight (g/mol) 166.151
MDL Number MFCD06202344
InChI Key HRPULQFHSZKTNA-UHFFFAOYSA-N
PubChem CID 14653132
IUPAC Name 7-fluoro-2,3-dihydrochromen-4-one
SMILES C1COC2=C(C1=O)C=CC(=C2)F

Specifications

Melting Point 46°C to 50°C
Quantity 250 mg
Synonym 7-fluorochroman-4-one, 7-fluoro-4-chromanone, 7-fluoro-3,4-dihydro-2h-1-benzopyran-4-one, 7-fluoro-2,3-dihydro-1-benzopyran-4-one, 7-fluoro-2,3-dihydro-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one,7-fluoro-2,3-dihydro, 4h-1-benzopyran-4-one, 7-fluoro-2,3-dihydro, pubchem17924, acmc-1bnvw, 7-fluoro-chroman-4-one
Formula Weight 166.15
Percent Purity 98%
Assay may contain up to 15% 5-isomer
Chemical Name or Material 7-Fluorochroman-4-one, sum of isomers
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.