missing translation for 'onlineSavingsMsg'
Learn More

7-Aminoindole, 97%, Thermo Scientific Chemicals

Product Code. 11364765
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5192-04-1
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.17
MDL Number MFCD02093960
InChI Key WTFWZOSMUGZKNZ-UHFFFAOYSA-N
Synonym 7-aminoindole, 7-amino-1h-indole, 1h-indol-7-ylamine, 7-indolamine, indole-7-ylamine, 7-indolylamine, 7-aminoindol, acmc-1ay6v, indol-7-yl amine, 7-aminoindole hplc
PubChem CID 111634
ChEBI CHEBI:33068
IUPAC Name 1H-indol-7-amine
SMILES C1=CC2=C(C(=C1)N)NC=C2

Specifications

Melting Point 90°C to 91°C
Quantity 100 mg
Beilstein 471787
Sensitivity Air sensitive
Formula Weight 132.17
Percent Purity 97%
Chemical Name or Material 7-Aminoindole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'