Learn More
YSI™ 626250 ProODO Cable and Probe
Stores calibration data within the sensor, allowing probes to be placed on any ProODO instrument without re-calibration
Brand: YSI™ 62625050
This item is not returnable.
View return policy
Description
For use with ProODO Handheld Meters exclusively.
- Assemblies include a ProODO optical DO sensor and sensing cap
- Cable assemblies over one meter also include a cable management kit
Specifications
±1%mg/L | |
50 m | |
RoHS, CE, WEEE, IP65 | |
0.1°C | |
DO/temperature meter | |
Stores calibration data within the sensor |
164 ft. | |
10°C to 40°C | |
Sensing cap and cable management kit | |
2 Years | |
ProODO |