Learn More
YSI™ 60520 Pro Series DO Cable
Compatible with the YSI Pro Plus or Pro20 handhelds
Brand:  YSI™ 605201
 View more versions of this product
Product Code. 11987117
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
Easy user-replaceable sensor allows for quick sensor replacements, if needed. No tools necessary. One-year warranty.Measure pH quickly and easily with the 1001 sensor.The 605101 pH Sensor can be used with the YSI Professional Plus Multiparameter Handheld Instrument. Requires Cable/Probe Assembly.Item is heat packed in the winter months. Shipment must be made via 2-day or next day carrier during that time information.
Cables over 1-meter also come with the 603062 cable management kit.
Order Info
Cables do NOT come with the sensor
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cable Assembly
3.2 ft.
1 Year
Measure temperature and DO
1 m