Learn More
YSI™ 60520 Pro Series DO Cable
Compatible with the YSI Pro Plus or Pro20 handhelds
Brand: YSI™ 60520100
This item is not returnable.
View return policy
Description
Easy user-replaceable sensor allows for quick sensor replacements, if needed. No tools necessary. One-year warranty.Measure pH quickly and easily with the 1001 sensor.The 605101 pH Sensor can be used with the YSI Professional Plus Multiparameter Handheld Instrument. Requires Cable/Probe Assembly.Item is heat packed in the winter months. Shipment must be made via 2-day or next day carrier during that time information.
Cables over 1-meter also come with the 603062 cable management kit.
Order Info
Cables do NOT come with the sensor
Specifications
Cable Assembly | |
328 ft. | |
2 Years |
Measure temperature and DO | |
100 m |