Learn More
3M™ 6000 Series Half Face Mask Respirator
Half-face mask respirators 6000 Series are economical, low-maintenance, and extremely lightweight. Probed reusable respirator assembly 6000Q series are used for quantitative fit purposes only.
Brand:  3M™ 7000146847
Product Code. 11753353
  • 24.84€ / Each
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Reusable half masks are economical, low-maintenance, simple-to-handle, and extremely lightweight
  • Used with 3M™ Supplied Air Systems
  • Bayonet Connection System allows connection to a broad range of twin lightweight filters to protect against gases, vapors, and particulates
  • Twin filter design provides lower breathing resistance
  • Probed reusable respirator assembly 6000 Series used for quantitative fit testing (17-987-3A & -3B)
Compliance
NIOSH approved under 42 CFR 84 standard for use against a variety of respiratory hazards when used as designed with appropriate cartridges and filters; in Canada, CSA standard Z94.4 and applicable local government requirements must be met for approval.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Gray
Half-Mask Respirator
K6200
Thermoplastic Elastomer
Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, painting, remediation, sanding, welding
4-point
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products