Learn More
3M™ 6000 Series Full Face Mask Respirators
Offers lightweight comfort and ease of use. 3M™ 6000 Series Reusable Full Face Mask Respirators have the 3M Bayonet Connection System, enabling connection to a broad range of twin lightweight filters to protect against gases, vapours and particulates.
Brand:  3M™ 7100015974
Product Code. 12589767
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • Large lens for wide field of view and excellent visibility
  • Well-balanced design with silicone face seal for greater comfort and durability
  • Unique center adapter directs exhaled breath and moisture downward, reducing valve debris and facilitating quick and easy cleaning
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EN 136:1998 (Class 1)
Light Gray
Reusable Full Face Mask Respirator
Full
200 series filters and 600 series cartridges
Bayonet connection, double gasket, phonic membrane, four-point head strap
Polycarbonate
400 g
Bayonet
Silicone, Thermoplastic Elastomer
Small
For construction applications
4-point
Polycarbonate Visor
1 Ea.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products