missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+/-)-6-Methyl-5-hepten-2-ol, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
(±)-6-Methyl-5-hepten-2-ol is a compound used in the synthesis of aggregation pheromone of the ambrosia beetle. It is also used as pharmaceutical intermediates and insect attractant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(+/-)-6-Methyl-5-hepten-2-ol is a compound used in the synthesis of aggregation pheromone of the ambrosia beetle. It is also used as pharmaceutical intermediates and insect attractant.
Solubility
Soluble in Chloroform. Not miscible with water.
Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
(+/-)-6-Methyl-5-hepten-2-ol is a compound used in the synthesis of aggregation pheromone of the ambrosia beetle. It is also used as pharmaceutical intermediates and insect attractant.
Solubility
Soluble in Chloroform. Not miscible with water.
Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Density | 0.84 |
Boiling Point | 78°C to 80°C (15 mmHg) |
Flash Point | 67°C (152°F) |
Refractive Index | 1.448 |
Quantity | 5 g |
Beilstein | 1720073 |
Solubility Information | Soluble in Chloroform. Not miscible with water. |
Formula Weight | 128.22 |
Percent Purity | 98% |
Chemical Name or Material | (+/-)-6-Methyl-5-hepten-2-ol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'