missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy-2H-chromene-3-carboxaldehyde, 97%, Thermo Scientific™

Product Code. 11383666
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134822-76-7
Molecular Formula C10H8O3
Molecular Weight (g/mol) 176.171
MDL Number MFCD01076587
InChI Key JSPGOHYLKVWWKI-UHFFFAOYSA-N
Synonym 6-hydroxychromene-3-carboxaldehyde, acmc-20alym, 6-hydroxy-2h-3-chromenecarbaldehyde, 6-hydroxy-2h-1-benzopyran-3-carboxaldehyde, 2h-1-benzopyran-3-carboxaldehyde, 6-hydroxy
PubChem CID 4770815
IUPAC Name 6-hydroxy-2H-chromene-3-carbaldehyde
SMILES C1C(=CC2=C(O1)C=CC(=C2)O)C=O

Specifications

Melting Point 164°C to 166°C
Quantity 1 g
Beilstein 6646155
Sensitivity Air Sensitive
Formula Weight 176.17
Percent Purity 97%
Chemical Name or Material 6-Hydroxy-2H-chromene-3-carboxaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'