Learn More
(5R,6S)-5,6-Diphenyl-2-morpholinone, 98%, Thermo Scientific™
Product Code. 15416828
Click to view available options
Quantity:
1 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
CAS 282735-66-4
Molecular Formula C16H15NO2
MDL Number MFCD09751026
Quantity 1 g
Synonym 5r,6s-5,6-diphenyl-2-morpholinone, 5r,6s-5,6-diphenylmorpholin-2-one, 5r, 6s-5, 6-diphenyl-2-morpholinone, 2-morpholinone, 5,6-diphenyl-, 5r,6s, pubchem18125, 2,3-diphenyl-morpholin-2-one, 5alpha,6alpha-diphenylmorpholine-2-one, 5r,6s-5,6-diphenyl-4-morpholin-2-one, 5r,6s-tetrahydro-5,6-diphenyl-2h-1,4-oxazin-2-one, 2s,3r-2,3,5,6-tetrahydro-2,3-diphenyl-1,4-oxazin-6-one
InChI Key LTPOSIZJPSDSIL-UHFFFAOYNA-N
SMILES O=C1CNC(C(O1)C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name 5,6-diphenylmorpholin-2-one
Molecular Weight (g/mol) 253.30
PubChem CID 11322747
Formula Weight 253.3
Percent Purity 98%
Physical Form Solid
Chemical Name or Material (5R,6S)-5,6-Diphenyl-2-morpholinone