missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ 5mm O.D. Thin Wall Precision NMR Sample Tube

Features 15% more sample volume than the standard wall Precision tube, and 25% more sample volume than the Economy NMR tube.

Brand:  Bel-Art™ 66605-0581

Product Code. 15379018

  • 65.15€ / Each
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

NMR tubes are thin glass walled tubes used to contain samples in nuclear magnetic resonance spectroscopy. It is important that the tubes are uniformly thick and well-balanced to ensure that they spin at regular rates (i.e., they do not wobble), within the NMR spectrometer. NMR tubes are used primarily with small samples dissolved in an appropriate solvent.

Specifications

Specifications

NMR Sample Tube
5 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions