Learn More
5-Chloro-2-iodoaniline, 98%, Thermo Scientific Chemicals
Product Code. 11389919
Click to view available options
Quantity:
1 g
5 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6828-35-9
Molecular Formula C6H5ClIN
Molecular Weight (g/mol) 253.467
MDL Number MFCD06738960
InChI Key FEOMAFDDLHSVMO-UHFFFAOYSA-N
Synonym benzenamine, 5-chloro-2-iodo, 5-chloro-2-iodo-phenylamine, 5-chloro-2-iodo-benzenamine, 5-chloro-2-iodophenylamine, 2-amino-4-chloro-iodobenzene, pubchem5286, 2-iodo-5-chloroaniline, 5-chloro-2-iodoanilline, 5-chloro-2-iodo-aniline, acmc-209o2s
PubChem CID 639697
IUPAC Name 5-chloro-2-iodoaniline
SMILES C1=CC(=C(C=C1Cl)N)I
Specifications
Melting Point 40°C to 42°C
Quantity 1 g
UN Number UN2811
Sensitivity Light sensitive
Formula Weight 253.47
Percent Purity 98%
Chemical Name or Material 5-Chloro-2-iodoaniline