missing translation for 'onlineSavingsMsg'
Learn More

5-Carboxamidotryptamine maleate, Tocris Bioscience™

Product Code. 30001091
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

5-HT1 agonist; also has high affinity for 5-ht5a and 5-HT7

5-HT1 agonist with high affinity at 5-HT1A, 5-HT1B, 5-HT1D, 5-ht5 and 5-HT7 receptors.

Chemical Identifiers

CAS 74885-72-6
Molecular Formula C11H13N3O·C4H4O4

Specifications

Quantity 50 mg
Formula Weight 319.32
Percent Purity >99%
Chemical Name or Material 5-Carboxamidotryptamine maleate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.