missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methoxy-beta-nitrostyrene, 97%, Thermo Scientific™

Product Code. 11486586
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175205-14-8
Molecular Formula C9H8BrNO3
Molecular Weight (g/mol) 258.071
MDL Number MFCD00068087
InChI Key IAHCHTBHTXVKTC-SNAWJCMRSA-N
Synonym 4-bromo-1-methoxy-2-2-nitrovinyl benzene, 4-bromo-1-methoxy-2-e-2-nitroethenyl benzene, 5-bromo-2-methoxy-omega-nitrostyrene, 2-1e-2-nitrovinyl-4-bromo-1-methoxybenzene, benzene,4-bromo-1-methoxy-2-2-nitroethenyl, beta-nitro-2-methoxy-5-bromostyrene
PubChem CID 5358770
IUPAC Name 4-bromo-1-methoxy-2-[(E)-2-nitroethenyl]benzene
SMILES COC1=C(C=C(C=C1)Br)C=C[N+](=O)[O-]

Specifications

Melting Point 120°C to 122°C
Quantity 1 g
Formula Weight 258.07
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-methoxy-beta-nitrostyrene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'