Learn More
Tocris Bioscience™ 5-BrdU
Synthetic thymidine analog; used for cell proliferation assays
Brand:  Tocris Bioscience™ 5015/50
Product Code. 18727467
  • 96.00€ / 50mg
Explore more special offers
Description
Description
5-BrdU is a synthetic thymidine analog; incorporated into DNA during replication. Used in assays for cell proliferation. Replaces Oct-4 in transcription factor-mediated reprogramming of somatic cells and can be used in small molecule cocktail to generate ciPSCs.
Specifications
Specifications
>99%
5-BrdU
5-BrdU
50 mg
307.1