missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-bromobenzoic acid, 95%, Thermo Scientific Chemicals

Product Code. 15416848
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and is also used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and is also used in medicine.

Solubility
Slightly soluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2840-02-0
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.03
MDL Number MFCD07368968
InChI Key FEXDUVBQBNYSQV-UHFFFAOYSA-N
Synonym 5-amino-2-bromo-benzoic acid, 4-bromo-3-carboxyaniline, benzoic acid,5-amino-2-bromo, benzoic acid, 5-amino-2-bromo, pubchem4637, acmc-209h2e, 5-amino-2-bromobenzoicacid, 2-bromo-5-aminobenzoic acid, ksc561g5h, 5-amino-2-bromo benzoic acid
PubChem CID 2769624
IUPAC Name 5-amino-2-bromobenzoic acid
SMILES NC1=CC(C(O)=O)=C(Br)C=C1

Specifications

Melting Point 171°C to 179°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 216.03
Percent Purity 95%
Chemical Name or Material 5-Amino-2-bromobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'