Learn More
5-Acetyl-2-thiophenecarbaldehyde, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Specifications
Specifications
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Quantity | 250 mg |
| Formula Weight | 154.19 |
| Percent Purity | 97% |
| Chemical Name or Material | 5-Acetyl-2-thiophenecarbaldehyde |