missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 5(6)-Carboxyfluorescein, mixture of isomers, 97%

Product Code. 11343067
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72088-94-9
Molecular Formula C42H24O14
Molecular Weight (g/mol) 752.64
MDL Number MFCD00151081
InChI Key BPVHBBXCESDRKW-UHFFFAOYSA-N
Synonym 5 6-fam, 3',6'-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9'-xanthene-5-carboxylic acid compound with 3',6'-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9'-xanthene-6-carboxylic acid 1:1, 5-and-6-fam, 5-and-6-carboxyfluorescein, 5-carboxyfluorescein; carboxyfluorescein, 5 6-carboxyfluorescein, dye content 90 %, 5 6-carboxyfluorescein; carboxyfluorescein, 5 6-carboxyfluorescein, bioreagent, suitable for fluorescence hplc, 3,6-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9-xanthene-5-carboxylic acid compound with 3,6-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9-xanthene-6-carboxylic acid 1:1, 4 5-carboxyfluorescein; 3',6'-dihydroxy-3-oxospiro 2-benzofuran-1,9'-xanthene-5 6-carboxylic acid
PubChem CID 44119975
IUPAC Name 3',6'-dihydroxy-1-oxospiro[2-benzofuran-3,9'-xanthene]-5-carboxylic acid;3',6'-dihydroxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-5-carboxylic acid
SMILES OC(=O)C1=CC2=C(C=C1)C1(OC2=O)C2=C(OC3=C1C=CC(O)=C3)C=C(O)C=C2.OC(=O)C1=CC2=C(C=C1)C(=O)OC21C2=C(OC3=C1C=CC(O)=C3)C=C(O)C=C2

Specifications

Melting Point ∼300°C
Quantity 1 g
Formula Weight 376.32
Percent Purity 97%
Chemical Name or Material 5(6)-Carboxyfluorescein, mixture of isomers

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'