Learn More
Tuttnauer 4th Dosing Pump WD Pump with Flow
Optional 4th dosing pump, for lubricant or rinse aid with flow meter.
Brand:  Tuttnauer 119904314
Product Code. 17808269
  • 595.00€ / 1 unit
Explore more special offers
Description
Description
  • Tiva Washer Accessory
  • 4th Dosing Pump WD Pump With flow
  • Dos. 4 Disinfectant/additive WD pump with flowmeter, for proper product dosage
Specifications
Specifications
Stainless Steel
Pump
Tiva Washer
1 Ea.