missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Labs Spectra/Por™ 6 3500 D MWCO Standard RC Pre-wetted Dialysis Kits

With Pre-wetted ready-to-use tubing, not requiring soaking to remove glycerol

162.00€ - 181.00€

Specifications

MWCO 3.5 kDa
Quantity 1 m Tubing/Kit
View More Specs

Includes:
  • 1 meter of Spectra/Por 6 Dialysis Tubing
  • 1 Standard Spectra/Por Closure (for buoyancy)
  • 1 Weighted Spectra/Por Closure (for orientation)
  • 5 Tubing Opening Picks

Products 3
Product Code Brand Diameter (Metric) Length (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Length (Metric) Price Quantity & Availability  
15370732
View Documents
Spectrum™
132592T
29 mm 6.4 mL/cm vol./length
181.00€
1m
Please to purchase this item. Need a web account? Register with us today!
15360732
View Documents
Spectrum™
132590T
11.5 mm 1.1 mL/cm vol./length
162.00€
1m
Please to purchase this item. Need a web account? Register with us today!
15380732
View Documents
Spectrum™
132594T
34 mm 9.3 mL/cm vol./length
180.00€
1m
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Clear, flexible, study standard grade Regenerated Cellulose (RC) dialysis membrane
  • Membrane packaged wet in preservative solution of 0.05% sodium azide
  • Kits available in 1m tubing lengths ideal for membrane evaluation and qualification
  • Include a pair of closures and convenient tubing opening picks

Specifications

Specifications

3.5 kDa
1 m Tubing/Kit
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.