missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4000 NTU Ratio Turbidity Standard, Reagecon
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
Suspension of extremely accurate (± 1%) polymer microspheres with guaranteed shelf life stability even after opening the bottle.
185.00€ - 384.00€
Specifications
For Use With (Equipment) | Ratio Instruments |
---|---|
Accuracy | Accuracy ± 1% NTU |
Product Code | Brand | Description | Volume (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Volume (Metric) | Price | Quantity & Availability | ||||
11556593
![]() |
Reagecon
CRSR-4000-100 |
Liquid format in an HDPE Bottle | 100 mL |
185.00€
100mL |
Estimated Shipment: 12-03-2025
Log in to see stock. |
||||
11566593
![]() |
Reagecon
CRSR-4000-500 |
Liquid format in an HDPE Bottle GL45 | 500 mL |
384.00€
500mL |
Estimated Shipment: 12-03-2025
Log in to see stock. |
||||
Description
- Turbidity Reference Standard, Turbidity Reference Material.
- Designed and formulated to overcome the adverse safety, accuracy and stability issues experienced with other turbidity standards, primarily formazin-based standards.
- Certified
- NIST traceable
- Non-toxic, non-carcinogenic
- Highly accurate
- Stable
- EPA approved
Specifications
Ratio Instruments | |
Accuracy ± 1% NTU |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title