missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 4-Phenylsemicarbazide, 98+%
Click to view available options
Quantity:
10 g
25 g
Specifications
Specifications
Melting Point | 123°C to 125°C |
Quantity | 25 g |
Beilstein | 511107 |
Sensitivity | Air Sensitive |
Merck Index | 14,7312 |
Formula Weight | 151.17 |
Percent Purity | ≥98% |
Chemical Name or Material | 4-Phenylsemicarbazide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'