missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-phenylbutyl isothiocyanate, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
Specifications
Specifications
| CAS Min % | 95.0 |
| Assay Percent Range | 95% |
| Packaging | Amber Glass Bottle |
| Quantity | 10 g |
| Formula Weight | 191.3 |
| Percent Purity | 95% |
| Chemical Name or Material | 4-Phenylbutyl isothiocyanate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido