missing translation for 'onlineSavingsMsg'
Learn More

4-Oxo-TEMPO, free radical, 96%, Thermo Scientific Chemicals

61.51€ - 202.01€

Chemical Identifiers

CAS 2896-70-0
Molecular Formula C9H16NO2
MDL Number MFCD00006187
Synonym 4-Oxo-2,2,6,6-tetramethyl-1-piperidinyloxy, free radical
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11384444
SDS View Documents
Thermo Scientific Alfa Aesar
L01084.03
1 g
61.51€
1g
Please to purchase this item. Need a web account? Register with us today!
11394444
SDS View Documents
Thermo Scientific Alfa Aesar
L01084.06
5 g
202.01€
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

4-Oxo-Tempo, Free radical is used as a useful Spin Trap for in Vitro Experiments. Reaction Between Tempone And Biologically Significant Reactive Oxygen Species can be monitored By Esr Spectroscopy. It is also used In the synthesis And Reactions Of Stable Nitroxyl Radicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Oxo-Tempo, Free radical is used as a useful Spin Trap for in Vitro Experiments. Reaction Between Tempone And Biologically Significant Reactive Oxygen Species can be monitored By Esr Spectroscopy. It is also used In the synthesis And Reactions Of Stable Nitroxyl Radicals.

Solubility
Soluble in water (partly), acetone, DMSO, 100% ethanol, and diethylether. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications

Chemical Identifiers

2896-70-0
C9H16NO2
MFCD00006187
4-Oxo-2,2,6,6-tetramethyl-1-piperidinyloxy, free radical

Specifications

2896-70-0
C9H16NO2
1818579
Soluble in water (partly),acetone,DMSO,100% ethanol,and diethylether. Insoluble in water.
96%
36°C to 40°C
MFCD00006187
4-Oxo-2,2,6,6-tetramethyl-1-piperidinyloxy, free radical
170.23
4-Oxo-TEMPO, free radical
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

missing translation for 'einecsNumber' : 220-778-7

missing translation for 'rtecsNumber' : TN9200000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Oxo-TEMPO, free radical, 96%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.