missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrophenyl-beta-D-glucuronide, Serva Electrophoresis™
Click to view available options
Quantity:
1 g
Specifications
CAS | 10344-94-2 |
Melting Point | 119°C to 122°C |
Color | White |
Molecular Formula | C12H12NO9 |
MDL Number | MFCD00036210 |
Quantity | 1 g |
Synonym | 4-nitrophenyl-beta-d-glucuronide, 4-nitrophenylglucuronide, 4-nitrophenyl beta-d-glucopyranosiduronic acid, 4-nitrophenyl beta-d-glucuronide, p-nitrophenyl beta-d-glucopyranosiduronic acid, 4-nitrophenyl b-d-glucosiduronic acid, p-nitrophenyl-b-d-glucuronide, 4-nitrophenyl b-d-glucuronide, p-nitrophenyl beta-d-glucuronide, beta-d-glucopyranosiduronic acid, 4-nitrophenyl, labeled with carbon-14 |
Solubility Information | Soluble in water |
InChI Key | QSUILVWOWLUOEU-GOVZDWNOSA-M |
SMILES | O[C@@H]1[C@@H](O)[C@H](OC2=CC=C(C=C2)[N+]([O-])=O)O[C@@H]([C@H]1O)C([O-])=O |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'