Learn More
BTX™ 4-needle Array Electrodes for BTX™ AgilePulse™ In Vivo
4-needle Array Electrodes are miniature needles designed to provide superior, highly-uniform electric fields in dermal or muscular tissue as part of the AgilePulse™ In Vivo Gene Delivery Electroporation System. Specially designed for gene and vaccine delivery.
396.00€ - 528.00€
Specifications
For Use With (Application) | Gene and Vaccine Delivery |
---|---|
For Use With (Equipment) | AgilePulse™ in Vivo Gene Delivery Electroporation System |
Material | Stainless Steel |
Product Line | BTX Electroporation |
Product Type | Electroporation Needle Array |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
15477370
|
BTX™
470040 |
2 mm |
396.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15487370
|
BTX™
470043 |
3 mm |
528.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15497370
|
BTX™
470045 |
5 mm |
528.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The miniature parallel-needle array is inserted directly into the target site for fast, reliable in vivo electroporation. The electric fields produced are the closest approximation of parallel plate electrodes, treating approximately 80% of the target area with 95% of the applied electric field.
The needle array requires the needle array handle which includes the electrode connector cable. Several needle lengths and row spacing are available for various intra-dermal or intra-muscular applications. Needle Arrays can be used for up to 500 pulses with proper care and maintenance.
- Uniform, reliable electric fields
- Miniature needles minimize tissue trauma
- Disposable and sterile
- Medical-grade plastic and surgical steel construction
- Safety-assured design
- Multiple configurations, sizes to choose from
Specifications
Gene and Vaccine Delivery | |
Stainless Steel | |
Electroporation Needle Array | |
0.3 mm | |
1 Ea. | |
4 mm | |
Sterile | |
Square Wave |
AgilePulse™ in Vivo Gene Delivery Electroporation System | |
BTX Electroporation | |
0.01 in. | |
4-needle Array | |
0.16 in. | |
Fine Point | |
Electrode Connector Cable |
Intended for Research use only. Not for use in diagnostic or clinical procedures.