missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-methoxybenzonitrile, 98%, Thermo Scientific Chemicals

Product Code. 11489196
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4421-08-3
MDL Number MFCD00001820

Specifications

Melting Point 85°C to 89°C
Molecular Formula C8H7NO2
Quantity 5 g
UN Number UN3439
Synonym vanillonitrile, m-anisonitrile, 4-hydroxy, 4-hydroxy-3-methoxy-benzonitrile, 3-methoxy-4-hydroxybenzonitrile, 4-hydroxy-3-methoxybenzenecarbonitrile, pubchem13112, 2-methoxy-4-cyanophenol, 4-cyano-2-methoxyphenol, 5-cyano-2-hydroxyanisole, 2-methoxy-4-cyano-phenol
InChI Key QJRWLNLUIAJTAD-UHFFFAOYSA-N
SMILES COC1=C(C=CC(=C1)C#N)O
IUPAC Name 4-hydroxy-3-methoxybenzonitrile
Molecular Weight (g/mol) 149.15
PubChem CID 78135
Formula Weight 149.15
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-3-methoxybenzonitrile
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'