missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-trans-beta-styrylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Product Code. 15497765
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 504433-86-7
Molecular Formula C14H18BFO2
Molecular Weight (g/mol) 248.10
MDL Number MFCD12546189
InChI Key ZJRAXVMUDOVAOD-UHFFFAOYSA-N
Synonym e-2-4-fluorostyryl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-fluoro-trans-beta-styrylboronic acid pinacol ester, 2-e-2-4-fluorophenyl ethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-p-fluorostyryl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-e-4-fluorostyryl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-trans-2-4-fluorophenyl vinyl-4,4,5,5-tetramethyl 1,3,2 dioxaborolane
PubChem CID 11207378
IUPAC Name 2-[(E)-2-(4-fluorophenyl)ethenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C=CC1=CC=C(F)C=C1

Specifications

Quantity 1 g
Formula Weight 248.11
Percent Purity 97%
Chemical Name or Material 4-Fluoro-trans-beta-styrylboronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'