missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-(trifluoromethoxy)aniline, 97%, Thermo Scientific™

Product Code. 15455577
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15455577

Brand: Thermo Scientific Alfa Aesar H32810.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Fluoro-3-(trifluoromethyl)aniline is used in the synthesis of trifluoromethylated poly(phenylene oxide). It is used in the preparation of 2-[3-(4-fluoro-3-trifluoromethyl-phenyl)-ureido]-2-methyl-propionic acid methyl ester by reacting with 2-isocyanato-2-methyl-propionic acid methyl ester. It is also used as a chemical intermediate in the synthesis of agrochemicals, pharmaceuticals and dyes.

Notes
Store in a cool place. Incompatible with strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113421-98-0
Molecular Formula C7H5F4NO
Molecular Weight (g/mol) 195.12
MDL Number MFCD09832246
InChI Key JSMGZPOPIVVIRZ-UHFFFAOYSA-N
Synonym 4-fluoro-3-trifluoromethoxy aniline, benzenamine, 4-fluoro-3-trifluoromethoxy, 4-fluoro-3-trifluoromethoxy-aniline, 4-fluoro-3-trifluoromethoxy phenylamine, 4-fluoro-3-trifluoro methoxy aniline
PubChem CID 21614378
IUPAC Name 4-fluoro-3-(trifluoromethoxy)aniline
SMILES NC1=CC(OC(F)(F)F)=C(F)C=C1

Specifications

Refractive Index 1.4502
Quantity 250 mg
UN Number UN2810
Formula Weight 195.12
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-(trifluoromethoxy)aniline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.