missing translation for 'onlineSavingsMsg'
Learn More

4-(Cyclohexyliminomethyl)benzeneboronic acid pinacol ester, Thermo Scientific™

Product Code. 15459175
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1218790-50-1
Molecular Formula C19H28BNO2
Molecular Weight (g/mol) 313.248
MDL Number MFCD11113042
InChI Key BAVKTLABAAKQAH-UHFFFAOYSA-N
Synonym 4-cyclohexyliminomethyl benzeneboronic acid pinacol ester, 1e-n-cyclohexyl-1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanimine, amtb116, 4-cyclohexyl iminomethylphenylboronic acid pinacol ester, 4-cyclohexyl iminomethyl phenyl boronic acid pinacol ester, e-n-cyclohexyl-1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanimine, 4-cyclohexyliminomethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzene, n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzylidene cyclohexanamine, n-cyclohexyl-1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanimine
PubChem CID 46739736
IUPAC Name N-cyclohexyl-1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanimine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C=NC3CCCCC3

Specifications

Quantity 1 g
Formula Weight 313.24
Chemical Name or Material 4-(Cyclohexyliminomethyl)benzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'