missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-N-Fmoc-L-phenylalanine, 95%, Thermo Scientific™

Product Code. 15488577
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 173963-93-4
Molecular Formula C25H20N2O4
Molecular Weight (g/mol) 412.445
MDL Number MFCD00672559
InChI Key JOPKKUTWCGYCDA-QHCPKHFHSA-N
Synonym fmoc-phe 4-cn-oh, fmoc-l-4-cyanophenylalanine, fmoc-dl-4-cyanophenylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-cyanophenyl propanoic acid, fmoc-l-4-cyanophe, fmoc-d-4-cyanophenylalanine, fmoc-4-cyano-l-phenylalanine, l-phenylalanine, 4-cyano-n-9h-fluoren-9-ylmethoxy carbonyl, 2s-3-4-cyanophenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-l-4-cn-phe-oh
PubChem CID 6957993
IUPAC Name (2S)-3-(4-cyanophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)C#N)C(=O)O

Specifications

Melting Point 190°C to 192°C
Quantity 1 g
Beilstein 7500424
Formula Weight 412.44
Percent Purity 95%
Chemical Name or Material 4-Cyano-N-Fmoc-L-phenylalanine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'