Learn More
4-Cyano-N-Fmoc-L-phenylalanine, 95%, Thermo Scientific™
Product Code. 15408587
Click to view available options
Quantity:
250 mg
1 g
5 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 173963-93-4
Molecular Formula C25H20N2O4
Molecular Weight (g/mol) 412.445
MDL Number MFCD00672559
InChI Key JOPKKUTWCGYCDA-QHCPKHFHSA-N
Synonym fmoc-phe 4-cn-oh, fmoc-l-4-cyanophenylalanine, fmoc-dl-4-cyanophenylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-cyanophenyl propanoic acid, fmoc-l-4-cyanophe, fmoc-d-4-cyanophenylalanine, fmoc-4-cyano-l-phenylalanine, l-phenylalanine, 4-cyano-n-9h-fluoren-9-ylmethoxy carbonyl, 2s-3-4-cyanophenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-l-4-cn-phe-oh
PubChem CID 6957993
IUPAC Name (2S)-3-(4-cyanophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)C#N)C(=O)O
Specifications
Melting Point 190°C to 192°C
Quantity 250 mg
Beilstein 7500424
Formula Weight 412.44
Percent Purity 95%
Chemical Name or Material 4-Cyano-N-Fmoc-L-phenylalanine