missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-iodoaniline, 98%, Thermo Scientific Chemicals

Product Code. 11401050
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63069-48-7
Molecular Formula C6H5ClIN
Molecular Weight (g/mol) 253.467
MDL Number MFCD01863737
InChI Key FLEJOBRWKBPUOX-UHFFFAOYSA-N
Synonym 2-iodo-4-chloroaniline, 4-chloro-2-iodo-aniline, 4-chloro-2-iodo-phenylamine, 4-chloro-2-iodophenylamine, benzenamine, 4-chloro-2-iodo, zlchem 532, pubchem3645, 4-chloro-6-iodoaniline, acmc-1bcyk, 2-iodo-4-chloro-aniline
PubChem CID 2757615
IUPAC Name 4-chloro-2-iodoaniline
SMILES C1=CC(=C(C=C1Cl)I)N

Specifications

Melting Point 41°C to 44°C
Flash Point >110(230°F)
Quantity 5 g
UN Number UN2811
Sensitivity Light sensitive
Formula Weight 253.47
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-iodoaniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'