Learn More
Applied Biosystems™ 3130/3100-Avant Genetic Analyzer 4-Capillary Array, 50 cm
Use for fragment analysis and sequencing. Usage per array is 100 runs.
Brand: Applied Biosystems™ 4333466
This item is not returnable.
View return policy
Description
This is a 4-capillary array (internally uncoated) for the 3130 and 3100 Avant™ Genetic Analyzer.Order Info
Each
Specifications
Store the array at ambient temperature. For details about storing the capillary array on and off the instrument, please refer to your genetic analyzer's maintenance, troubleshooting and reference guide. We guarantee the performance of the 3130 Capillary arrays up to 100 runs and within 1 year of purchase. |
|
50 cm | |
3100-Avant Genetic Analyzer, 3130 Genetic Analyzer | |
165 min | |
Room Temperature |
3130XL/3100 Genetic Analyzer 16-Capillary Array, 50cm Capillary Length, 61cm x 50μm I.D. Capillary, 165 min. Run Time, Use for Short Fragment Analysis, Room Temperature | |
Sequencing, Fragment Analysis | |
Avant™ | |
Genetic Analyzer |